missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
S1P3/EDG-3 Monoclonal antibody specifically detects S1P3/EDG-3 in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | S1P3/EDG-3 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | SR1721 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human S1P3/EDG-3 (Uniprot #: Q99500) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?