missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Recombinant Monoclonal [Clone S100A1/1706R] Rabbit, Clone: S100A1/1706R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 6271-RBM6-P1
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
S100A1 Recombinant Monoclonal [Clone S100A1/1706R] | |
Cytoplasmic | |
S100A1/1706R | |
Unconjugated | |
S100A1 | |
Recombinant full-length human S100A1 protein | |
Brain, Melanoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
Rabbit | |
10-12kDa | |
Protein A/G Purified | |
RUO | |
6271 | |
IgG κ |