Learn More
S100A1 Antibody (049), Alexa Fluor™ 532 , Novus Biologicals™
Código de producto. 18415429 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
18415429 0.1 mL 0.10 ml
1 options
Código de producto. 18415429 Proveedor Novus Biologicals N.º de proveedor NBP290504AF532
Rabbit Monoclonal Antibody
S100A1 Monoclonal antibody specifically detects S100A1 in Mouse samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno S100A1
Aplicaciones ELISA
Clasificación Monoclonal
Clon 049
Conjugado Alexa Fluor 532
Formulación 50mM Sodium Borate
Alias de gen protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Especie del huésped Rabbit
Inmunógeno This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A1 (P56565; Gly2-Ser94).
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 6271
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG