Learn More
RWDD4A Antibody, Novus Biologicals™
Código de producto. p-200038037 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25ul
Tamaño de la unidad:
0.10 ml
25 microlitros
18479550 25ul 25 microlitros
18222528 0.1 mL 0.10 ml
2 options
Código de producto. 18479550
Marca: Novus Biologicals NBP18474325ul
Rabbit Polyclonal Antibody
RWDD4A Polyclonal specifically detects RWDD4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RWDD4A
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen FAM28A, member A, MGC10198, Protein FAM28A, RWD domain containing 4, RWD domain containing 4A, RWD domain-containing protein 4, RWD domain-containing protein 4A, RWDD4A
Símbolos de los genes RWDD4
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:EALRSIYEGDESFRELSPVSFQYRIGENGDPKAFLIEISWTETYPQTPPILSMNAFFNNTISSAVKQSILAKLQEAVEANLGTAM
Método de purificación Affinity Purified
Cantidad 25ul
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 201965
Especificidad de la prueba Specificity of human RWDD4A antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only