Learn More
Eurofins DiscoverX Rsk2/MAPKAP Kinase 1b Protein, Active
Descripción
Rsk2, also known as Ribosomal Protein S6 Kinase A3, is a member of the RSK family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in this gene have been associated with Coffin-Lowry syndrome (CLS).
The protein sequence is identical to that described inGenBank NM_004586 for human RSK2, over residues 2-end. The cDNA however is more closely related to the mouse sequence described inGenBank AY083469.
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecular | 85 kDa |
| Cantidad | 10 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | N-terminal 6His |
| Sistema de expresión | Recombinant enzyme expressed in Sf21 insect cells |
| Forma de proteína | Truncated |
| Grado de pureza o calidad | ≥71% |
| Expresión | RSK2, active |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.