missing translation for 'onlineSavingsMsg'
Learn More

RSK1, p Ser380 Antibody (1E11), Novus Biologicals™

Código de producto. p-200022766 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
50 μL
Unit Size:
100 microlitros
50 microlitros
Product Code. Cantidad unitSize
30034195 50 μL 50 microlitros
30029325 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 30034195

Brand: Novus Biologicals NBP32639550ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RSK1 Monoclonal antibody specifically detects RSK1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antígeno RSK1
Aplicaciones Western Blot, ELISA, Immunoprecipitation
Clasificación Monoclonal
Clon 1E11
Conjugado Unconjugated
Dilución Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000
Formulación PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gen 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from Human RSK1 [p Ser380] [UniProt Q15418]
Método de purificación Affinity purified
Cantidad 50 μL
Estado normativo RUO
Disciplina de investigación MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 6195
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.