Learn More
RPE65 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL488-17939-20UL
Descripción
The RPE65 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPE65. This antibody recognizes human, mouse, rat antigen. The RPE65 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
| RPE65 | |
| 1000 μg/mL | |
| LCA2, mRPE65, rd12, Retinoid isomerohydrolase, Retinol isomerase, RP20, RPE65, sRPE65 | |
| 20 μL | |
| 6121 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |