Learn More
RP105/CD180 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18530434 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
25 microgramos
18530434 0.025 mg 25 microgramos
1 options
Código de producto. 18530434
Marca: Novus Biologicals NBP1767050.025mg
Rabbit Polyclonal Antibody
RP105/CD180 Polyclonal specifically detects RP105/CD180 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RP105/CD180
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen BAA12019
Alias de gen CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Símbolos de los genes CD180
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human RP105. The immunogen is located within amino acids 590 - 640 of RP105. Amino Acid Squence: NPPSLRGVKLSDVK
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 4064
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only