missing translation for 'onlineSavingsMsg'
Learn More

RNMTL1 Antibody - Azide and BSA Free, Novus Biologicals™

Código de producto. p-200060969 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.02 ml
0.10 ml
Código de producto. Cantidad unitSize
18637260 0.02 mL 0.02 ml
18616180 0.1 mL 0.10 ml
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18637260

Brand: Novus Biologicals NBP2932800.02ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNMTL1 Polyclonal antibody specifically detects RNMTL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno RNMTL1
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000
Formulación PBS (pH 7.3), 50% glycerol
Alias de gen EC 2.1.1, EC 2.1.1.-, FLJ10581, HC90, putative RNA methyltransferase, RNA methyltransferase like 1, RNA methyltransferase-like protein 1
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-420 of human MRM3 (NP_060616.1). MAALVRPARFVVRPLLQVVQAWDLDARRWVRALRRSPVKVVFPSGEVVEQKRAPGKQPRKAPSEASAQEQREKQPLEESASRAPSTWEESGLRYDKAYPGDRRLSSVMTIVKSRPFREKQGKILLEGRRLISDALKAGAVPKMFFFSRLEYLKELPVDKLKGVSLIKVKFEDIKDWSDLVTPQGIMGIFAKPDHVKMTYPKTQLQHSLPLLLICDNLRDPGNLGTILRSAAGAGCSKVLLTKGCVDAWEPKVLRAGMGAHFRMPIINNLEWETVPNYLPPDTRVYVADNCGLYAQAEMSNKASDHGWVCDQRVMKFHKYEEEEDVETGASQDWLPHVEVQSYDSDWTEAPAAVVIGGETYGVSLESLQLAESTGGKRLLIPVVPGVDSLNSAMAASILLFEGKRQLRGRAEDLSRDRSYH
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación DNA replication Transcription Translation and Splicing
Primario o secundario Primary
ID de gen (Entrez) 55178
Especies diana Human
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.