Learn More
RNF8 Mouse, anti-Human, Clone: 3G3H5, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 68521-1-IG-20UL
 Ver más versiones de este producto
Código de producto. 18029825
  • 145.00€ / 20 microlitros
Descripción
The RNF8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RNF8. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The RNF8 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
RNF8
3G3H5
FLJ12013, KIAA0646, ring finger protein 8, RNF8
20 μL
9025
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Rabbit, Pig
150 kDa
IgG1