Learn More
RNF160 Mouse, anti-Human, Clone: 1D2A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68610-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The RNF160 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RNF160. This antibody recognizes human, rat, mouse antigen. The RNF160 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| RNF160 | |
| 1D2A2 | |
| C21orf10, C21orf98, FLJ11053, KIAA0714, ring finger protein 160, RNF160, Zinc finger protein 294, ZNF294 | |
| 150 μL | |
| 26046 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG1 |