Learn More
RNF160 Mouse, anti-Human, Clone: 1D2A2, Proteintech
Código de producto. p-200050045 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18035515 20 μL 20 microlitros
18090536 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18035515 Proveedor Proteintech N.º de proveedor 686101IG20UL
Mouse Monoclonal Antibody
The RNF160 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RNF160. This antibody recognizes human, rat, mouse antigen. The RNF160 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Antígeno RNF160
Aplicaciones Western Blot, ELISA
Clon 1D2A2
Concentración 1000 μg/mL
Símbolos de los genes C21orf10, C21orf98, FLJ11053, KIAA0714, ring finger protein 160, RNF160, Zinc finger protein 294, ZNF294
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 26046
Especies diana Human, Rat, Mouse
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG1