missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Rit2 Monoclonal specifically detects Rit2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones
Especificaciones
| Antígeno | Rit2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI4A6 |
| Conjugado | DyLight 650 |
| Dilución | Western Blot |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2 |
| Símbolos de los genes | RIT2 |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?