Learn More
RIPK3/RIP3 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18715903 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 miligramo
25 microgramos
Cantidad:
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. unitSize Cantidad
18715903 25 microgramos -
18796103 0.10 miligramo 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18715903 Proveedor Novus Biologicals N.º de proveedor NBP224588SS
Rabbit Polyclonal Antibody has been used in 23 publications
RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Aplicaciones Western Blot, Immunohistochemistry