missing translation for 'onlineSavingsMsg'
Learn More

RHEBL1 Antibody, Novus Biologicals™

Código de producto. p-200066437 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18634835 100 μg 100 microlitros
18645115 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18634835 Proveedor Novus Biologicals N.º de proveedor NBP299023100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

RHEBL1 Polyclonal antibody specifically detects RHEBL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno RHEBL1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1:200-1:1000
Formulación PBS (pH 7.0)
Alias de gen FLJ25797, GTPase RhebL1, MGC34869, Ras homolog enriched in brain like 1, Ras homolog enriched in brain like 1 c, Ras homolog enriched in brain like-1 c, Ras homolog enriched in brain-like protein 1, Rheb2, RHEBL1c, Rheb-like protein 1
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human RHEBL1.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 121268
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.