missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10385-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Especificaciones
| RFC5 | |
| Polyclonal | |
| Unconjugated | |
| RFC5 | |
| A1 36 kDa subunit, Activator 1 36 kDa subunit, Activator 1 subunit 5, Replication factor C subunit 5, RF C 36 kDa subunit, RFC36, RFC5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304528, 5985, 72151 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P40937, Q9D0F6 | |
| RFC5 | |
| RFC5 Fusion Protein Ag0584 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction