missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | Ret |
| Aplicaciones | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | RET/2663 |
| Conjugado | Alexa Fluor 750 |
| Dilución | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence |
| Símbolos de los genes | RET |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?