missing translation for 'onlineSavingsMsg'
Learn More

RelA/NFkB p65 Antibody (112A1021) - BSA Free, Novus Biologicals™

Código de producto. 18708433 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
Código de producto. Cantidad unitSize
18708433 0.1 mg 0.10 miligramo
18069993 0.025 mg 25 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18708433

Marca: Novus Biologicals NB10056712

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 13 publications

RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno RelA/NFkB p65
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Clasificación Monoclonal
Clon 112A1021
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 2 - 5 ug/mL. Use reported in scientific literature (Wittmann et al (2005)), Simple Western 1:25. Use reported by customer review, Flow Cytometry 0.25 - 0.5 ug/10^6 cells, Immunohistochemistry 5 - 15 ug/mL, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 5 ug/mL. Use reported in scientific literature (Chang et al (2005)), Flow (Intracellular) reported in scientific literature (Wittmann et al (2005)), CyTOF-ready
N.º de referencia del gen Q969V5
Alias de gen anti-NF kB p65, anti-NFkB p65, anti-NF-kB p65, anti-p65, anti-RelA, anti-rela p65, MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65 flow cytometry, p65 immunohistochemistry, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Símbolos de los genes RELA
Especie del huésped Mouse
Inmunógeno This NfkB p65 antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Método de purificación Protein G purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 5970
Especificidad de la prueba Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.