missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 GST-Papain-like Protease, CF
Description
Recombinant SARS-CoV-2 virus Papain-like protease (PLPro) is a Ubiquitin- and ISG15-deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial PLPro concentration of 20-100 nM when using Ubiquitin-Rhodamine 110 (U-555) substrate. When using polyubiquitin chain substrate(s), this enzyme demonstrates a preference for K48 linkages.
Specifications
Specifications
| Número de acceso | YP_009725299.1 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | Supplied as a solution in HEPES, NaCl, Glycerol and TCEP. |
| Cantidad | 50 μg |
| Fuente | E. coli-derived sars-cov-2 Papain-like Protease protein Glu746 - Lys1060, with an N-terminal GST-tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugado | Unconjugated |
| Grado de pureza o calidad | >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
| Expresión | Papain-like Protease |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?