Learn More
R&D Systems™ Recombinant SARS-CoV-2 BA.2 Spike RBD Fc Chimera Protein
Descripción
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Especificaciones
Especificaciones
| Número de acceso | YP_009724390.1 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Peso molecular | M.W.-observed: 59-65 kDa, under reducing conditions., M.W.-theroretical: 52 kDa |
| Cantidad | 100 μg |
| Fuente | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein SARS-CoV-2 BA0.2 Spike RBD (Arg319-Phe541) (Gly339Asp, Ser371Phe, Ser373Pro, Ser375Phe, Thr376Ala, Asp405Asn, Arg408Ser, Lys417Asn, Asn440Lys, Ser477Asn, Thr478Lys, Glu484Ala, Gln493Arg, Gln498Arg, Asn501Tyr, Tyr505His) (C-terminus) Accession # YP_009724390.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentración de endotoxinas | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugado | Unconjugated |
| Reconstitución | Reconstitute at 500 μg/mL in PBS. |
| Mostrar más |