missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Tnf alpha Protein

Código de producto. 15986279
Change view
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
500 μg
5 μg
Unit Size:
1 miligramo
100 microgramos
20 microgramos
5 microgramos
500 microgramos
Product Code. Cantidad unitSize
15986279 20 μg 20 microgramos
15906289 5 μg 5 microgramos
15966279 100 μg 100 microgramos
15996279 500 μg 500 microgramos
15976279 1 mg 1 miligramo
5 options
This item is not returnable. View return policy

Product Code. 15986279

Brand: enQuireBio™ QP550820ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Specifications

Número de acceso NP_036807.1
ID de gen (Entrez) 103694380
Nombre Tnf alpha Protein
Cantidad 20 μg
Estado normativo Research Use Only
Concentración de endotoxinas Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Símbolo de gen Tnf
Actividad biológica Cytolysis of mouse L929 cells in the presence of Actinomycin D significantly induced by less than 50 pg/ml corresponding to an activity of at least 2.0 x 10^7 units/mg (or a typical ED50 of less than 10 pg/ml).
Tipo de producto Recombinant Protein
Reactividad cruzada Rat
Especie E. coli
Etiqueta de proteína Untagged
Secuencia MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL
Tampón 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Grado de pureza o calidad Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.