Learn More
enQuireBio™ Recombinant Rat Rantes Protein
Especificaciones
Especificaciones
| ID de gen (Entrez) | 81780 |
| Nombre | Rantes Protein |
| Cantidad | 5 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | < 1.0 EU per ug protein as determined by the LAL method. |
| Símbolo de gen | Ccl5 |
| Actividad biológica | The activity is determined by the ability to chemoattract total human lymphocytes and murine T-cells at a concentration between 1-10ng/ml corresponding to a Specific Activity of 100,000-1,000,000IU/mg. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Rat |
| Especie | E. coli |
| Mostrar más |