Learn More
R&D Systems™ Recombinant Rat Lipocalin-2/NGAL Protein
Description
The Recombinant Rat Lipocalin-2/NGAL Protein is derived from NS0. The Recombinant Rat Lipocalin-2/NGAL Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulación | Lyophilized from a 0.2μm filtered solution in MES and NaCl. |
| ID de gen (Entrez) | 170496 |
| Peso molecular | 21 kDa |
| Nombre | Lipocalin-2/NGAL |
| Cantidad | 50 μg |
| Fuente | Mouse myeloma cell line,NS0-derived rat Lipocalin-2/NGAL protein Gln21-Asn198,with a C-terminal 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Alias de gen | 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Show More |