Learn More
R&D Systems™ Recombinant Rat IL-5 Protein
Descripción
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.
Especificaciones
Especificaciones
| Formulación | Liofilizado a partir de una solución filtrada de 0,2 μm en PBS con BSA como proteína transportadora. |
| ID de gen (Entrez) | 24497 |
| Peso molecular | 13 kDa (monomer) |
| Nombre | IL-5 |
| Cantidad | 10 μg |
| Fuente | Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-5 protein Met20-Val132 |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forma estructural | Disulfide-linked homodimer |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Mostrar más |