missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine TNF a Protein
A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10884-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
397086 | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
MSTESMIRDV ELAEEALAKK AGGPQGSRRC LCLSLFSFLL VAGATTLFCL LHFEVIGPQK EEFPAGPLSI NPLAQGLRSS SQTSDKPVAH VVANVKAEGQ LQWQSGYANA LLANGVKLKD NQLVVPTDGL YLIYSQVLFR GQGCPSTNVF LTHTISRIAV SYQTKVNLLS AIKSPCQRET PEGAEAKPWY EPIYLGGVFQ LEKDDRLSAE INLPDYLDFA ESGQVYFGII AL |
TNF a Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by the cytotoxicity of porcine PK15 cells was found to be 0.001-0.015 ng/ml. | |
Porcine | |
Untagged | |
The protein was lyophilized with no additives. |