Learn More
enQuireBio™ Recombinant Porcine TNF a Protein
Código de producto. p-7148695
Click to view available options
Cantidad:
1 mg
20 μg
5 μg
Tamaño de la unidad:
1 miligramo
20 microgramos
5 microgramos
Código de producto. 15915468
Marca: enQuireBio™ QP108845ug
A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 397086
Nombre TNF a Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Símbolo de gen TNF
Actividad biológica The ED50 as determined by the cytotoxicity of porcine PK15 cells was found to be 0.001-0.015 ng/ml.
Tipo de producto Recombinant Protein
Reactividad cruzada Porcine
Especie E. coli
Etiqueta de proteína Untagged
Secuencia MSTESMIRDV ELAEEALAKK AGGPQGSRRC LCLSLFSFLL VAGATTLFCL LHFEVIGPQK EEFPAGPLSI NPLAQGLRSS SQTSDKPVAH VVANVKAEGQ LQWQSGYANA LLANGVKLKD NQLVVPTDGL YLIYSQVLFR GQGCPSTNVF LTHTISRIAV SYQTKVNLLS AIKSPCQRET PEGAEAKPWY EPIYLGGVFQ LEKDDRLSAE INLPDYLDFA ESGQVYFGII AL
Tampón The protein was lyophilized with no additives.
Grado de pureza o calidad Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.