missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Treponema TmpA 45kDa Protein

Código de producto. 15994779
Change view
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
500 microgramos
Código de producto. Cantidad unitSize
15994779 100 μg 100 microgramos
15914789 500 μg 500 microgramos
15904789 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 15994779

Brand: enQuireBio™ QP13812100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the Treponema TmpA 45kDa was constructed and used to recombinantly synthesize the protein.

Specifications

Nombre Treponema TmpA 45kDa Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Other
Especie E. coli
Etiqueta de proteína His
Tampón Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10.
Grado de pureza o calidad Greater than 95.0% as determined by SDS-PAGE.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.