Learn More
enQuireBio™ Recombinant other SPA-Cys His Protein
Especificaciones
Especificaciones
| Nombre | SPA-Cys His Protein |
| Cantidad | 100 mg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Símbolo de gen | spa |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Other |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Secuencia | MNAAQHDEAQ QNAFYQVLNM PNLNADQRNG FIQSLKDDPS QSANVLGEAQ KLNDSQAPKA DAQQNNFNKD QQSAFYEILN MPNLNEAQRN GFIQSLKDDP SQSTNVLGEA KKLNESQAPK ADNNFNKEQQ NAFYEILNMP NLNEEQRNGF IQSLKDDPSQ SANLLSEAKK LNESQAPKAD NKFNKEQQNA FYEILHLPNL NEEQRNGFIQ SLKDDPSQSA NLLAEAKKLN DAQAPKADNK FNKEQQNAFY EILHLPNLTE EQRNGFIQSL KDDPSVSKEI LAEAKKLNDA QAPKEEDSLE HHHHHHC |
| Mostrar más |