Learn More
enQuireBio™ Recombinant other Filamin Protein
Código de producto. p-7149732
Change view
Click to view available options
Cantidad:
10 μg
1 mg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
15998678 10 μg 10 microgramos
15918688 50 μg 50 microgramos
15908688 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 15998678 Proveedor enQuireBio™ N.º de proveedor QP1188310ug
A cDNA sequence encoding the Filamin was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre Filamin Protein
Cantidad 10 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Other
Especie Non-species Specific
Etiqueta de proteína Untagged
Tampón The protein was lyophilized from a 1 mg/ml solution containing 20mM Tris / acetate buffer pH 7.6, 0.1mM EDTA, 2mM DTT, 9M urea and 20mM NaCl.
Grado de pureza o calidad Greater than 90.0% as determined by SDS-PAGE.