missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Cyclosporin A Protein

A cDNA sequence encoding the Cyclosporin A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11582-4gram

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15997708

  • 189.00€ / 4 g
Fecha estimada de envío: 31-07-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

Cyclosporin A Protein
4 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 99.0% as determined by RP-HPLC.
Research Use Only
Recombinant Protein
Native Protein
The Cyclosporin-A was lyophilized from a concentrated (1 mg/ml) solution with no additives.
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales