missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Chimeric Chagas Protein
Specifications
Specifications
| Nombre | Chimeric Chagas Protein |
| Cantidad | 100 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Other |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Tampón | Chimeric Chagas Multiantigen was lyophilized from 20mM Tris pH 8.5, 100mM Sodium chloride, 20% trehalose and 0.1% Sodium azide as preservative. |
| Grado de pureza o calidad | Protein is >90% pure as determined by metal affinity chromatography. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?