Learn More
enQuireBio™ Recombinant Multi Species ACTH Protein
Código de producto. 15945668
Change view
Click to view available options
Cantidad:
2 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
2 miligramos
50 miligramos
15945668 2 mg 2 miligramos
15935668 10 mg 10 miligramos
15955668 50 mg 50 miligramos
3 options
Código de producto. 15945668
Marca: enQuireBio™ QP109502mg
Recombinant Protein
Especificaciones
Nombre Multi Species ACTH Protein
Cantidad 2 mg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Multi Species
Etiqueta de proteína Untagged
Secuencia Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH
Tampón The ACTH hormone was lyophilized with no additives.
Grado de pureza o calidad Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.