missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF Protein
Specifications
Specifications
| Nombre | VEGF Protein |
| Cantidad | 2 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | < 1.0 EU per ug protein as determined by the LAL method. |
| Actividad biológica | The ED50 range, determined by the dose-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (measured by 3H-thymidine uptake) is 1-2 ng/ml, corresponding to a specific activity of 1x106 Units/mg. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Mouse |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Secuencia | MNFLLSWVHW TLALLLYLHH AKWSQAAPTT EGEQKSHEVI KFMDVYQRSY CRPIETLVDI FQEYPDEIEY IFKPSCVPLM RCAGCCNDEA LECVPTSESN ITMQIMRIKP HQSQHIGEMS FLQHSRCECR PKKDRTKPEK KSVRGKGKGQ KRKRKKSRFK SWSVHCEPCS ERRKHLFVQD PQTCKCSCKN TDSRCKARQL ELNERTCRCD KPRR |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?