missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TREM2 Protein

Código de producto. 15975599
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
20 μg
500 μg
50 μg
Unit Size:
1 miligramo
10 microgramos
100 microgramos
20 microgramos
200 microgramos
50 microgramos
500 microgramos
Product Code. Cantidad unitSize
15975599 100 μg 100 microgramos
15995589 10 μg 10 microgramos
15925599 20 μg 20 microgramos
15985599 50 μg 50 microgramos
15915599 200 μg 200 microgramos
15935599 500 μg 500 microgramos
15905599 1 mg 1 miligramo
7 options
This item is not returnable. View return policy

Product Code. 15975599

Brand: enQuireBio™ QP4435ma100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) TREM2, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_001259007.1
ID de gen (Entrez) 83433
Nombre TREM2 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen Trem2
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Mouse
Especie Mammal
Etiqueta de proteína His
Secuencia LNTTVLQGMA GQSLRVSCTY DALKHWGRRK AWCRQLGEEG PCQRVVSTHG VWLLAFLKKR NGSTVIADDT LAGTVTITLK NLQAGDAGLY QCQSLRGREA EVLQKVLVEV LEDPLDDQDA GDLWVPEESS SFEGAQVEHS TSRNQETSFP PTS
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.