Learn More
R&D Systems™ Recombinant Mouse sTNF RI/TNFRSF1A Protein
Descripción
The Recombinant Mouse sTNF RI/TNFRSF1A Protein is derived from E. coli. The Recombinant Mouse sTNF RI/TNFRSF1A Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
| Formulación | Liofilizado a partir de una solución filtrada de 0,2 μm en PBS con BSA como proteína transportadora. |
| ID de gen (Entrez) | 21937 |
| Peso molecular | 21 kDa |
| Nombre | TNF RI/TNFRSF1A |
| Cantidad | 50 μg |
| Fuente | E. coli-derived mouse TNF RI/TNFRSF1A protein Ile22-Ala212,with an N-terminal Met |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Grado de pureza o calidad | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |