Learn More
R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus
Código de producto. 18715526
Click to view available options
Cantidad:
200 μg
25 μg
Formulación:
Liofilizado a partir de una solución filtrada de 0,2 μm en NaH2PO4 NaCl and DTT con BSA como proteína transportadora.
Liofilizado a partir de una solución filtrada de 0,2 μm en NaH2PO4 NaCl y DTT.
Tamaño de la unidad:
200 microgramos
25 microgramos
Código de producto. 18715526
Marca: R&D Systems™ 464SH200
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Especificaciones
Formulación Liofilizado a partir de una solución filtrada de 0,2 μm en NaH2PO4 NaCl and DTT con BSA como proteína transportadora.
ID de gen (Entrez) 20423
Peso molecular 19.8 kDa
Nombre Sonic Hedgehog/Shh (C25II), N-Terminus
Cantidad 200 μg
Fuente E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile),with an N-terminal Met
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Alias de gen HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugado Unconjugated
Recombinante Recombinant
Grado de pureza o calidad 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.