missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL5 Protein

Product Code. 15983908
Change view
Click to view available options
Cantidad:
100 μg
250 μg
25 μg
5 μg
Unit Size:
100 microgramos
25 microgramos
250 microgramos
5 microgramos
Product Code. Cantidad unitSize
15983908 100 μg 100 microgramos
15913918 5 μg 5 microgramos
15903918 25 μg 25 microgramos
15993908 250 μg 250 microgramos
4 options
This item is not returnable. View return policy

Product Code. 15983908

Brand: enQuireBio™ QP10454ec100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mouse IL5, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_034688.1
ID de gen (Entrez) 16191
Nombre IL5 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Less than 1.0 EU/ug as determined by LAL method.
Símbolo de gen IL5
Longitud de la proteína Partial (see sequence information for more details).
Actividad biológica The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Tipo de producto Recombinant Protein
Reactividad cruzada Mouse
Especie E. coli
Etiqueta de proteína Untagged
Secuencia MEIPMSTVVK ETLTQLSAHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEMLFQNL SLIKKYIDRQ KEKCGEERRR TRQFLDYLQE FLGVMSTEWA MEG
Tampón 0.2 m filtered 20 mM Tris, pH 9.0, 150 mM NaCl, lyophilized
Grado de pureza o calidad Greater than 98% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.