missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-17A Protein
Description
- Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.
- ED50 for this effect is 0.25 to 1.25 ng/mL.
Specifications
Specifications
| Número de acceso | Q62386.1 |
| Para utilizar con (aplicación) | Bioactivity |
| ID de gen (Entrez) | 3605 |
| Peso molecular | 15.5 kDa (monomer) |
| Nombre | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Cantidad | 100 μg |
| Fuente | E. coli-derived mouse IL-17/IL-17A protein Thr22-Ala158 |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Símbolo de gen | Il17a |
| Conjugado | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?