missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-12AB Protein
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
2 μg
500 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
500 microgramos
Especificaciones
Especificaciones
| Número de acceso | NP_001290173.1 |
| ID de gen (Entrez) | 16160 |
| Nombre | IL-12AB Protein |
| Cantidad | 10 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Símbolo de gen | IL12 |
| Actividad biológica | Shows bioactivity at a concentration of 20 ng/mL in the Mouse primary T cell induced expression of CD25 assay. The bioactivity acceptance criteria is set at 5 x 10^4 units/mg. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Mouse |
| Mostrar más |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction