missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cytochrome P450 2E1 Protein

Código de producto. 16017641
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16017641 200 μg 200 microgramos
16097631 10 μg 10 microgramos
16037641 50 μg 50 microgramos
16087631 100 μg 100 microgramos
16027641 500 μg 500 microgramos
16007641 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16017641 Supplier enQuireBio™ Supplier No. QP9094ye200ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 2E1, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_067257.1
ID de gen (Entrez) 13106
Nombre Cytochrome P450 2E1 Protein
Cantidad 200 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen Cyp2e1
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Mouse
Especie Yeast
Etiqueta de proteína His
Secuencia AVLGITVALL VWIATLLLVS IWKQIYRSWN LPPGPFPIPF FGNIFQLDLK DIPKSLTKLA KRFGPVFTLH LGQRRIVVLH GYKAVKEVLL NHKNEFSGRG DIPVFQEYKN KGIIFNNGPT WKDVRRFSLS ILRDWGMGKQ GNEARIQREA HFLVEELKKT KGQPFDPTFL IGCAPCNVIA DILFNKRFDY DDKKCLELMS LFNENFYLLS TPWIQAYNYF SDYLQYLPGS HRKVMKNVSE IRQYTLGKAK EHLKSLDINC PRDVTDCLLI EMEKEKHSQE PMYTMENISV TLADLFFAGT ETTSTTLRYG LLILMKYPEI EEKLHEEIDR VIGPSRAPAV RDRMNMPYMD AVVHEIQRFI NLVPSNLPHE ATRDTVFRGY VIPKGTVVIP TLDSLLFDNY EFPDPETFKP EHFLNENGKF KYSDYFKAFS AGKRVCVGEG LARMELFLLL SAILQHFNLK SLVDPKDIDL SPVTIGFGSI PREFKLCVIP RS
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.