missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H7N7 Netherlands Protein
A cDNA sequence encoding the H7N7 Netherlands was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12095-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza H7N7 Netherlands Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
The Recombinant H7N7 A/Netherlands/219/03 solution contains 10mM Sodium phosphate, pH 7 and 150mM NaCl. |