missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Influenza A Virus H1N1 Hemagglutinin Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human Galectin-1 (Catalog # 1152-GA ) immobilized at 10.00 μg/mL, 100 μL/well, the concentration of Recombinant Influenza A Virus H1N1 HA Tag His-tag (Catalog # 10973-HA) that produces 50% of the optimal binding response is 0.8-8.0 μg/mL.
Specifications
Specifications
| Número de acceso | YP_009118626.1 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Peso molecular | MolecularWeight-observed: 68-83 kDa, under reducing conditions., MolecularWeight-theroretical: 58 kDa |
| Cantidad | 100 μg |
| Fuente | Human embryonic kidney cell, HEK293-derived influenza a virus h1n1 Hemagglutinin protein Asp18-Gln529, with a C-terminal 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentración de endotoxinas | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugado | Unconjugated |
| Reconstitución | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?