missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UPF0454 protein C12orf49 Protein

Código de producto. 16002601
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
missing translation for 'unitSize'
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
This item is not returnable. View return policy

Product Code. 16002601

missing translation for 'mfr': enQuireBio™ QP8088ec100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) UPF0454 protein C12orf49, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_079014.1
ID de gen (Entrez) 79794
Nombre UPF0454 protein C12orf49 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen C12orf49
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His-SUMO
Secuencia TFKQEERAVR DRNLLQVHDH NQPIPWKVQF NLGNSSRPSN QCRNSIQGKH LITDELGYVC ERKDLLVNGC CNVNVPSTKQ YCCDGCWPNG CCSAYEYCVS CCLQPNKQLL LERFLNRAAV AFQNLFMAVE DHFELCLAKC RTSSQSVQHE NTYRDPIAKY CYGESPPELF PA
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.