missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TPI1 Protein

Product Code. 15974719
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
25 μg
5 μg
missing translation for 'unitSize'
25 microgramos
5 microgramos
Product Code. Cantidad unitSize
15974719 25 μg 25 microgramos
15984719 5 μg 5 microgramos
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 15974719

missing translation for 'mfr': enQuireBio™ QP1379325ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the TPI1 was constructed and used to recombinantly synthesize the protein.

Specifications

ID de gen (Entrez) 7167
Nombre TPI1 Protein
Cantidad 25 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen TPI1
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MAPSRKFFVG GNWKMNGRKQ SLGELIGTLN AAKVPADTEV VCAPPTAYID FARQKLDPKI AVAAQNCYKV TNGAFTGEIS PGMIKDCGAT WVVLGHSERR HVFGESDELI GQKVAHALAE GLGVIACIGE KLDEREAGIT EKVVFEQTKV IADNVKDWSK VVLAYEPVWA IGTGKTATPQ QAQEVHEKLR GWLKSNVSDA VAQSTRIIYG GSVTGATCKE LASQPDVDGF LVGGASLKPE FVDIINAKQ
Tampón The TPI1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Grado de pureza o calidad Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.