missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombopoietin/TPO Protein, Animal-Free

Produktkod. p-200112495
Change view
Klicka för att se tillgängliga alternativ
Cantidad:
1 mg
50 μg
Förpackningsstorlek:
1 miligramo
50 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Cantidad unitSize
30306042 1 mg 1 miligramo
30306066 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30306042 Leverantör R&D Systems™ Leverantörsnummer BTTPOAFL01M

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.050-0.500 ng/mL. The specific activity of Recombinant Human Thrombopoietin is >1 x 107 units/mg, which is calibrated against the human Thrombopoietin reference standard (NIBSC code: 03/124).
TRUSTED_SUSTAINABILITY

Specifikationer

Número de acceso NP_000451.1
Para utilizar con (aplicación) Bioactivity
Formulación Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
ID de gen (Entrez) 7066
Peso molecular M.W.-theoretical: 19 kDa, M.W.-observed: 18-19 kDa, under reducing conditions.
Cantidad 1 mg
Fuente E. coli-derived human Thrombopoietin/Tpo protein Ser22-Leu195 Produced using non-animal reagents in an animal-free laboratory.
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Concentración de endotoxinas <0.10 EU per 1 μg of the protein by the LAL method.
Alias de gen Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Conjugado Unconjugated
Reconstitución Reconstitute at 500 μg/mL in water.
Grado de pureza o calidad Antigen Affinity-purified
Expresión Thrombopoietin/THPO
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.