Learn More
enQuireBio™ Recombinant Human TFF3 / Trefoil Factor 3 Protein
A DNA sequence encoding the Human TFF3/Trefoil Factor 3, was expressed in the hosts and tags indicated.
105.00€ - 1970.00€
Especificaciones
Número de acceso | NP_003217.3 |
---|---|
ID de gen (Entrez) | 7033 |
Nombre | TFF3 / Trefoil Factor 3 Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15972998
|
enQuireBio™
QP10180-ec-5ug |
5 μg | N/A | ||||||
15952998
|
enQuireBio™
QP10180-ec-20ug |
20 μg | N/A | ||||||
15942998
|
enQuireBio™
QP10180-ec-100ug |
100 μg | N/A | ||||||
15962998
|
enQuireBio™
QP10180-ec-250ug |
250 μg | N/A | ||||||
Especificaciones
NP_003217.3 | |
TFF3 / Trefoil Factor 3 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EEYVGLSANQ CAVPAKDRVD CGYPHVTPKE CNNRGCCFDS RIPGVPWCFK PLQEAECTF | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
7033 | |
Research Use Only | |
TFF3 | |
The ED50 as determined by a chemotaxis bioassay using human MCF-7 cells is less than 10 ug/ml, corresponding to a specific activity of > 100 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Título del producto
enQuireBio™ Recombinant Human TFF3 / Trefoil Factor 3 Protein