Learn More
enQuireBio™ Recombinant Human SURF1 Protein
Código de producto. 15904279
Change view
Click to view available options
Cantidad:
1 mg
25 μg
5 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
5 microgramos
15904279 5 μg 5 microgramos
15994269 25 μg 25 microgramos
15984269 1 mg 1 miligramo
3 options
Código de producto. 15904279
Marca: enQuireBio™ QP136515ug
A cDNA sequence encoding the SURF1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre SURF1 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MQVQRRKWKL NLIAELESRV LAEPVPLPAD PMELKNLEYR PVKVRGCFDH SKELYMMPRT MVDPVREARE GGLISSSTQS GAYVVTPFHC TDLGVTILVN RGFVPRKKVN PETRQKGQIE GEVDLIGMVR LTETRQPFVP ENNPERNHWH YRDLEAMARI TGAEPIFIDA NFQSTVPGGP IGGQTRVTLR NEHLQ
Tampón The Surfeit-1 solution contains 20 mM Tris-HCl buffer (pH 8.0), 0.1M Nacl, 1mM DTT and 20% glycerol.
Grado de pureza o calidad Greater than 95.0% as determined by SDS-PAGE.