Learn More
enQuireBio™ Recombinant Human SORD Protein
Código de producto. p-7148678
Click to view available options
Cantidad:
1 mg
25 μg
5 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
5 microgramos
Código de producto. 15905418
Marca: enQuireBio™ QP108685ug
A cDNA sequence encoding the SORD was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 6652
Nombre SORD Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Símbolo de gen SORD
Actividad biológica Specific activity is > 0.2 units/mg, in which one unit will convert 1.0 umole of D-fructose to D-sorbitol per minute at pH 7.5 at 25C.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MAAAAKPNNL SLVVHGPGDL RLENYPIPEP GPNEVLLRMH SVGICGSDVH YWEYGRIGNF IVKKPMVLGH EASGTVEKVG SSVKHLKPGD RVAIEPGAPR ENDEFCKMGR YNLSPSIFFC ATPPDDGNLC RFYKHNAAFC YKLPDNVTFE EGALIEPLSV GIHACRRGGV TLGHKVLVCG AGPIGMVTLL VAKAMGAAQV VVTDLSATRL SKAKEIGADL VLQISKESPQ EIARKVEGQL GCKPEVTIEC TGAEASIQAG IYATRSGGTL VLVGLGSEMT TVPLLHAAIR EVDIKGVFRY CNTWPVAISM LASKSVNVKP LVTHRFPLEK ALEAFETFKK GLGLKIMLKC DPSDQNP
Tampón SORD protein solution (0.5 mg/ml) is formulated in 20mM Tris-HCl pH 8.0, 0.2M NaCl, 5mM DTT and 20% glycerol.
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.