Learn More
enQuireBio™ Recombinant Human RB1 Protein
Código de producto. 15973049
Change view
Click to view available options
Cantidad:
10 μg
1 mg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
50 microgramos
15973049 10 μg 10 microgramos
15993049 50 μg 50 microgramos
15983049 1 mg 1 miligramo
3 options
Código de producto. 15973049
Marca: enQuireBio™ QP1327510ug
A cDNA sequence encoding the RB1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre RB1 Protein
Cantidad 10 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MASFPSSPLRIPGGNIYISPLKSPYKISEGLPTPTKMTPRSRILVSIGESFGTSEKFQKINQMVCNSDRVLKRSAEGSNPPKPLKKLRFDIEGSDEADGSKHLPGESKFQQKLAEMTSTRTRMQKQKMNDSMDTSNKEEKHHHHHH
Tampón The RB1 (1 mg/ml) was lyophilized after extensive dialyses against 1xPBS pH 7.4.
Grado de pureza o calidad Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.