missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RARRES2 / Chemerin Protein

Código de producto. 15995939
Change view
Click to view available options
Cantidad:
100 μg
1 mg
25 μg
500 μg
5 μg
Unit Size:
1 miligramo
100 microgramos
25 microgramos
5 microgramos
500 microgramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15995939 1 mg 1 miligramo
15925949 5 μg 5 microgramos
15905949 25 μg 25 microgramos
15985939 100 μg 100 microgramos
15915949 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15995939 Supplier enQuireBio™ Supplier No. QP53491mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Specifications

Número de acceso NP_002880.1
ID de gen (Entrez) 5919
Nombre RARRES2 / Chemerin Protein
Cantidad 1 mg
Estado normativo Research Use Only
Concentración de endotoxinas Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Símbolo de gen RARRES2
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína Untagged
Secuencia MELTEAQRRG LQVALEEFHK HPPVQWAFQE TSVESAVDTP FPAGIFVRLE FKLQQTSCRK RDWKKPECKV RPNGRKRKCL ACIKLGSEDK VLGRLVHCPI ETQVLREAEE HQETQCLRVQ RAGEDPHSFY FPGQFAFS
Tampón Sterile filtered (0.22 um) solution containing 0.1% TFA
Grado de pureza o calidad Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.